Learn More
Invitrogen™ Phospho-Axl (Tyr698) Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-Axl (Tyr698) |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | AXL |
| Gene Accession No. | P30530, Q00993 |
| Gene Alias | Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog |
| Gene Symbols | AXL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.