Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BAD (Ser112) Monoclonal Antibody (G.445.9)
GREENER_CHOICE

Catalog No. PIMA515085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515085 Supplier Invitrogen™ Supplier No. MA515085
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

The protein encoded by BAD is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BAD (Ser112)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone G.445.9
Concentration 66 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene BAD
Gene Accession No. O35147, Q61337, Q92934
Gene Alias AI325008; BAD; Bbc2; Bbc6; Bcl2 antagonist of cell death; BCL2 associated agonist of cell death; bcl-2 associated death agonist; Bcl2-antagonist of cell death; BCL2-antagonist of cell death protein; bcl2-associated agonist of cell death; bcl2-associated death promoter; bcl-2-binding component 6; BCL2-binding component 6; BCL2-binding protein; BCL2L8; bcl2-L-8; Bcl-2-like protein 8; BCL-X/BCL-2 binding protein; bcl-XL/Bcl-2-associated death promoter; hypothetical protein LOC615013; OTTMUSP00000017561; OTTMUSP00000022400
Gene Symbols BAD
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pSer112 of mouse Bad.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12015, 572, 64639
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.