Learn More
Invitrogen™ Phospho-beta Catenin/gamma Catenin (Tyr333, Tyr324) Polyclonal Antibody

Description
The antibody detects a 92kDa* protein corresponding to the molecular mass of beta-Catenin on SDS-PAGE immunoblots of human A431 cells treated with pervanadate, but not in control cells. In addition, the pervanadate induced reactivity is not observed after alkaline phosphatase treatment.
Specifications
Specifications
| Antigen | Phospho-beta Catenin/gamma Catenin (Tyr333, Tyr324) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | CTNNB1 |
| Gene Accession No. | P35222, Q02248, Q9WU82 |
| Gene Alias | Catenin beta1; CTNNB1 |
| Gene Symbols | CTNNB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.