Learn More
Invitrogen™ Phospho-ZFP36L1 (Ser92) Polyclonal Antibody

Description
A suggested positive control for Western blot is HepG2 cells.
Specifications
Specifications
| Antigen | Phospho-ZFP36L1 (Ser92) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZFP36L1 |
| Gene Accession No. | P23950, Q07352 |
| Gene Alias | AW742437; AW743212; Berg36; Brf1; butyrate response factor 1; cMG1; D530020L18Rik; early response factor Berg36; EGF-inducible protein CMG1; EGF-response factor 1; ERF1; ERF-1; mRNA decay activator protein ZFP36L1; RNF162B; TIS11b; TPA-induced sequence 11b; ZFP36 ring finger protein like 1; ZFP36 ring finger protein-like 1; ZFP36L1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1 |
| Gene Symbols | ZFP36L1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.