Learn More
Invitrogen™ Phospho-BRG1 (Ser610) Polyclonal Antibody

Description
Antibody detects endogenous levels of BRG1 only when phosphorylated at Ser610.
Specifications
Specifications
| Antigen | Phospho-BRG1 (Ser610) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Smarca4 |
| Gene Accession No. | P51532, Q3TKT4, Q8K1P7 |
| Gene Alias | ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; BRG1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; mitotic growth and transcription activator; MRD16; nuclear protein GRB1; protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; Snf2b; SNF2beta; SNF2-beta; SNF2L4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; transcription activator BRG1 |
| Gene Symbols | Smarca4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.