Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-C17orf96 (Ser15) Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188465
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3664 10 μL
50-199-3663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3664 Supplier Active Motif Supplier No. 61746
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Elongin BC and Polycomb repressive complex 2-associated protein (EPOP) also know as C17ORF96 is a scaffold protein that serves as a bridging partner between the PRC2/EED-EZH2 complex and the elongin BC complex. It is required to fine tune the transcriptional status of polycomb group (PcG) target genes in embryonic stem cells (ESCs). C17ORF96 also plays a key role in genomic regions that display both active and repressive chromatin properties in pluripotent stem cells by sustaining low level expression of PcG target genes. It acts by recruiting the elongin BC complex, thereby restricting excessive activity of the PRC2/EED-EZH2 complex. C17ORF96 also has interacts with USP7 and promotes deubiquitination of H2B at promoter sites.

Specifications

Antigen Phospho-C17orf96 (Ser15)
Applications Dot Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene EPOP
Gene Accession No. Q7TNS8
Gene Alias AA409164; AI413509; AW539173; C17orf96; C9H17orf96; chromosome 17 open reading frame 96; chromosome 9 open reading frame, human C17orf96; E13; E130012A19Rik; Elongin BC and Polycomb repressive complex 2-associated protein; Embryonic stem cell-specific PRC2 subunit p48; EPOP; ES cell-specific PRC2 subunit p48; esPRC2p48; hypothetical protein LOC691153; LOC691153; proline rich 28; Proline-rich protein 28; PRR28; RIKEN cDNA E130012A19 gene; uncharacterized protein C17orf96; uncharacterized protein C17orf96 homolog; Uncharacterized protein ENSP00000317905
Gene Symbols EPOP
Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide containing phospho-serine 15 of human C17orf96.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103551
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.