Learn More
Description
PA1-060 detects phospho-caveolin-2 Y19 from human and mouse samples. PA1-060 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~21 kDa protein representing phospho-caveolin-2 Y19 from extract from COS-7 cells transiently transfected with the mouse caveolin-2 gene. Immunofluorescent staining of phospho-caveolin-2 Y19 in NIH 3T3 cells using PA1-060 results in a distinct punctate staining pattern at the cell periphery. The PA1-060 immunogen is a synthetic phosphopeptide corresponding to residues M(14) A D D A (pY) S H H S G C(25) of mouse CAV2. This peptide (Cat. # PEP-180) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Phospho-Caveolin 2 (Tyr19) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | CAV2 |
| Gene Accession No. | P51636, Q9WVC3 |
| Gene Alias | AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 |
| Gene Symbols | CAV2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
