Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CDC2 (Tyr15) (CDK1) Polyclonal Antibody, Abbomax

Catalog No. 502204045
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4045 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4045 Supplier Abbomax Inc Supplier No. 602160
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: A431 cell lysate Cellular location: Nuclear.

Phospho-CDC2 (Tyr15) (CDK1) Polyclonal antibody specifically detects Phospho-CDC2 (Tyr15) (CDK1) in Bovine, Chicken, Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Phospho-CDC2 (Tyr15) (CDK1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene CDK1
Gene Accession No. P06493, P13863, P48734
Gene Alias 5363; ABL1; cdc; CDC2; cdc-2; cdc2 kinase; CDC28A; Cdc2a; cdc2-a; cdc2Dm; cdc2x1.1; CDCDm; CDK1; Cdk-1; cdk1.S; CDK1/CDC2; CDK1-A; cdk1-b; Cdk1-PA; CDKN1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; cell division control protein 2 homolog; Cell division control protein 2 homolog 1; cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 1-A; CG5363; CG5363-PA; cyclin dependent kinase; cyclin dependent kinase 1; cyclin-dependent kinase; Cyclin-dependent kinase 1; cyclin-dependent kinase 1 S homeolog; cyclin-dependent kinase 1-A; D2366; Dcdc2; dCdk1; DKFZp686L20222; Dm cdc2; Dmcdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; DNA polymerase delta catalytic subunit; DNA polymerase III; DNA-directed DNA polymerase delta POL3; group 4; HPR6; KCIP-1; l(2)31Eh; LD38718p; maturation-promoting factor; MGC111195; MGC53829; MPF; OTTHUMP00000019660; P34 CDK1; P34 MGC111195; p34 protein kinase; p34 protein kinase 1; p34<CDC2>; p34cdc2; p34-cdc2 homologue; POL3; Protein 1054; protein 2; PSTAIR; TEX1; unnamed protein product; xcdc2; XELAEV_18036802mg; YDL102W
Host Species Rabbit
Immunogen A synthetic peptide surrounding to the epitope -GTYGV- with a phosphorylation site at Tyr15 of human CDC2 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281061, 396252, 983
Target Species Bovine, Chicken, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.