Learn More
Invitrogen™ Phospho-Cdc25B (Ser323) Polyclonal Antibody

Description
This antibody is predicted to react with human and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-Cdc25B (Ser323) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CDC25B |
| Gene Accession No. | P30305, P30306, P48966 |
| Gene Alias | AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 |
| Gene Symbols | CDC25B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.