Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CdGAP (Thr789) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564659 Supplier Invitrogen™ Supplier No. PA564659
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-CdGAP (Thr789) Polyclonal Antibody detects endogenous levels of CdGAP only when phosphorylated at Thr789.

This gene encodes a GTPase-activating protein (GAP). A variety of cellular processes are regulated by Rho GTPases which cycle between an inactive form bound to GDP and an active form bound to GTP. This cycling between inactive and active forms is regulated by guanine nucleotide exchange factors and GAPs. The encoded protein is a GAP shown to regulate two GTPases involved in protein trafficking and cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CdGAP (Thr789)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP31
Gene Accession No. A6X8Z5, Q2M1Z3
Gene Alias 5830477L08Rik; AOS1; ARHGAP31; AU041750; Cdc42 GTPase-activating protein; CDGAP; D330026I07Rik; KIAA1204; Rho GTPase activating protein 31; rho GTPase-activating protein 31
Gene Symbols ARHGAP31
Host Species Rabbit
Immunogen A synthesized peptide derived from human CdGAP (Phospho-Thr789).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12549, 288093, 57514
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.