Learn More
Invitrogen™ Phospho-CHK1 (Ser345) Recombinant Rabbit Monoclonal Antibody (PS01-17)

Description
Predicted band size: 54 kDa Positive Control: HeLa treated with Etoposide cell lysate, HeLa treated with Hydroxyurea cell lysate, HeLa treated with 4mM Hydroxyurea for 20 hours. Subcellular Location: Nucleus, Chromosome, Cytoplasm, cytoskeleton, microtubule organizing center, centrosome. Sequence Similarities: 100% Mouse/Rat. Tissue Specificity: Expressed ubiquitously with the most abundant expression in thymus, testis, small intestine and colon.
Specifications
Specifications
| Antigen | Phospho-CHK1 (Ser345) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | PS01-17 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide |
| Gene | Chek1 |
| Gene Accession No. | O14757 |
| Gene Alias | C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.