Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CHK2 (Thr68) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985605
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5605 Supplier Bioss Supplier No. BS3721R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Checkpoint kinase 2 (Chk2) is a serine/threonine-protein kinase which functions as a cell cycle checkpoint regulator. ATM and ATR kinases are activated in response to ionizing and ultraviolet radition, and phosphorylate Thr68 and other sites in the amino-terminal region of Chk2 dimerisation and activation. Activated Chk2 phosphorylates and inhibits Cdc25C phosphates, preventing the entry into mitosis, and has been shown to stabilize the tumor suppressor protein p53, leading to cell cycle arrest in G1. In addition, Chk2 can phosphorylate BRCA1, allowing BRCA1 to restore survival after DNA damage. Chk2 is a putative tumor suppressor protein, with mutations in to the Chk2 gene linked to Li-Fraumeni syndrome, a familiar cancer usually associated with inherited mutations in TP53. Also, mutations in this gene are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors.

Specifications

Antigen Phospho-CHK2 (Thr68)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Chek2
Gene Accession No. O96017
Gene Alias CDS1; Cds1 homolog; Checkpoint kinase 2; checkpoint-like protein CHK2; Chek2; CHK2; Chk2 (phospho T68); Chk2 (pThr68); CHK2 checkpoint homolog; Chk2 phospho; Chk2 phospho T68; del9; EC 2.7.11.1; hCds1; Hucds1; kinase Chk2; LFS2; OTTHUMP00000199044; OTTHUMP00000199045; OTTHUMP00000199064; OTTHUMP00000199115; OTTHUMP00000199116; phospho Chk2 (T68); phospho Thr68 Chk2; PP1425; protein kinase Chk2; RAD53; Rad53 homolog; RP11-436C9.1; Serine/threonine-protein kinase Chk2
Gene Symbols Chek2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human CHK2 around the phosphorylation site of Thr68.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11200
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.