Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Claspin (Ser30) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105980 Supplier Invitrogen™ Supplier No. PA5105980
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Claspin only when phosphorylated at Ser30.

Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Claspin (Ser30)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLSPN
Gene Accession No. Q80YR7, Q9HAW4
Gene Alias B130025E01; C85083; claspin; claspin homolog; CLSPN; E130314M08Rik; hClaspin; MGC131612; MGC131613; MGC131615
Gene Symbols CLSPN
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLSPN(Accession Q9HAW4), corresponding to amino acid residues around phosphorylated Ser30.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269582, 298534, 63967
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.