Learn More
Invitrogen™ Phospho- Connexin 36 (Ser276) Polyclonal Antibody

Description
Specific for endogenous levels of the ~35 kDa connexin-35/36 protein phosphorylated at Ser276.
Specifications
Specifications
| Antigen | Phospho- Connexin 36 (Ser276) |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | ∼0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM HEPES with 100 μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5 |
| Gene | GJD2 |
| Gene Accession No. | O54851 |
| Gene Alias | connexin 36; connexin36; connexin-36; cx36; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; Gja9; GJD2 |
| Gene Symbols | GJD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.