Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Cortactin (Tyr466) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 44856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-856 Supplier Invitrogen™ Supplier No. 44856
  • $500.00
    $375.65 / Each of 1

    Save $124.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Phospho-Cortactin (Tyr466) Polyclonal Antibody for Western Blot

Cortactin (Cttn) is a ubiquitous actin-binding protein that was originally identified as a substrate for Src. It contributes to the organization of the actin cytoskeleton and cell shape. Cortactin also plays a role in the formation of lamellipodia and cell migration. It is accumulated in peripheral, actin-enriched structures of cells suggesting that cortactin facilitates actin network formation. Cortactin has four major domains of interest the N-terminal acidic (NTA) and tandem repeats domains and the C-terminal proline-rich and SH3 Domains. NTA associates with the Arp2/3 and WASP complex at F-actin branches. Cortactin is involved in promoting cell motility and invasion, including a critical role in invadopodia, actin rich-subcellular protrusions associated with degradation of the ECM by cancer cells. Cortactin is phosphorylated by src family kinases at Y421, Y466, and Y482 and S405 and S418 that are phosphorylated by Erk family kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Cortactin (Tyr466)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene Cttn
Gene Accession No. Q60598
Gene Alias 1110020L01Rik; amplaxin; cortactin; Cttn; Cttnb; EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); FLJ34459; mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); oncogene EMS1; Oncogene SRC8; Src substrate cortactin; src8; Unknown (protein for MGC:137923)
Gene Symbols Cttn
Host Species Rabbit
Immunogen antiserum was produced against a chemically synthesized phosphopeptide derived from the region of mouse cortactin that contains tyrosine 466 (tyrosine 470 in the human sequence).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13043
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.