Learn More
Invitrogen™ Phospho-CrkL (Tyr207) Polyclonal Antibody

Description
Positive Control: K562, Neuro2A, NIH-3T3, C2C12, Rat2 Predicted Reactivity: Rhesus Monkey (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-CrkL (Tyr207) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.72 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | CRKL |
| Gene Accession No. | P46109, P47941, Q5U2U2 |
| Gene Alias | 1110025F07Rik; AA589403; AI325100; CRK like proto-oncogene, adaptor protein; CRKL; crkl {ECO:0000312; crk-like protein; crk-like protein {ECO:0000250; Crkol; EMBL:AAH85865.1, ECO:0000312; OTTHUMP00000198539; RGD:1308531}; UniProtKB:P46109}; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk sarcoma virus CT10 oncogene homolog-like |
| Gene Symbols | CRKL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.