Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-cytokeratin-8 (K8) (Ser73), Mouse anti-Mouse, Human, Clone: LJ4, Millipore Sigma™

Catalog No. MABT34425UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT34425UG 25 μg
MABT344100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT34425UG Supplier MilliporeSigma Supplier No. MABT34425UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-cytokeratin-8 (K8) (Ser73), clone LJ4, Cat. No. MABT344, is a mouse monoclonal antibody that detects human type II cytoskeletal 8 phophrorylated on serine 73 and murine cytokeratin-8 phophorylated on serine 79 and has been tested for use in ELISA, Immunocytochemistry, Immunofluoresce

Keratin, type II cytoskeletal 8 (UniProt: P05787; also known as Cytokeratin-8, CK-8 Keratin-8, K8, Type-II keratin Kb80) is encoded by the KRT8 (also known as CYK8) gene (Gene ID: 3856) in human. Cytokeratin-8 is a type II, neutral to basic, protein of the intermediate filament family that together with Cytokeratin-19 (KRT19) helps to link the contractile apparatus to dystrophin at the costameres of striated muscle. Cytokeratin-8 can undergo phosphorylation on three major serine residues: Serine 23, 431, and 73. Serine 23 is shown to be highly conserved in all type II keratins. Phosphorylation at Serine 73 is reported to increase during cellular stress, including hear and drug exposure. However, under normal conditions serine 73 remains largely dephosphorylated. Cytokeratin-8 can also undergo O-glycosylation in a cell cycle-dependent manner and glycosylation increases its solubility and reduces stability by inducing proteasomal degradation. Mutations in KRT8 gene have been linked to liver cirrhosis that is characterized by severe panlobular liver-cell swelling with Mallory body formation, prominent pericellular fibrosis, and marked deposits of copper. Two isoforms of Cytokeratin-8 have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-cytokeratin-8 (K8) (Ser73)
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone LJ4
Conjugate Unconjugated
Dilution Immunofluorescence Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser73) in Immunofluorescence applications (McIntosh, P.B., et. al. (2010). J Cell Sci. 123(Pt 16):2810-22; Liao, J., et. al. (1997). J Biol Chem. 272(28):17565-73; Ku, N.O., et. al. (2003). Hepatology. 37(5):1006-14; Toivola, D.M., et. al. (2002). Mol Biol Cell. 13(6):1857-70).Immunoprecipitation Analysis: A representative lot immunoprecipitaated phospho-cytokeratin-8 (K8) (Ser73) in Immunoprecipitation applications (Ku, N.O., et. al. (2003). Hepatology. 37(5):1006-14; Liao, J., et. al. (1997). J Biol Chem. 272(28):17565-73).Immunocytochemistry Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser73) in Immunocytochemistry applications (Toivola, D.M., et. al. (2002). Mol Biol Cell. 13(6):1857-70).Western Blotting Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser73) in Western Blotting applications (Toivola, D.M., et. al. (2002). Mol Biol Cell. 13(6):1857-70; Ku, N.O., et. al. (2003). Hepatology. 37(5):1006-14; Liao, J., et. al. (1997). J Biol Chem. 272(28):17565-73; McIntosh, P.B., et. al. (2010). J Cell Sci. 123(Pt 16):2810-22).ELISA Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser73) in ELISA applications (Liao, J., et. al. (1997). J Biol Chem. 272(28):17565-73).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Keratin;type II cytoskeletal 8;CK-8;Keratin-8;K8;Type-II keratin Kb80
Gene Symbols KRT8;CYK8
Host Species Mouse
Immunogen Cytokeratin-8 purified from HT29 cells treated with okadaic acid (1 mg/ml, 2h).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001243222
Test Specificity Clone LJ4 detects human cytokeratin-8 phosphorylated on serine 73 and murine cytokeratin-8 phosphorylated on serine 79.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.