Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 44530G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-530-G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-530-G Supplier Invitrogen™ Supplier No. 44530G
  • $552.00
    $414.65 / Each of 1

    Save $137.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody for Western Blot

This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-eIF2b epsilon (Ser539)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene Eif2b5
Gene Accession No. Q13144, Q64350, Q8CHW4
Gene Alias C81315; CACH; CLE; EIF-2B; eIF-2B GDP-GTP exchange factor subunit epsilon; EIF2B5; EIF2BE; EIF2Bepsilon; eukaryotic translation initiation factor 2B subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; LVWM; Translation initiation factor eIF-2B subunit epsilon
Gene Symbols Eif2b5
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of rat eIF2Be that contains serine 535 (corresponding to serine 539 in the human sequence).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192234, 224045, 8893
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.