Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho Enol Pyruvate Carboxylase Polyclonal Antibody, Biotin, Rockland™
SDP

Catalog No. 501991189
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1189 25 μL
50-199-1188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1189 Supplier Rockland Immunochemicals Supplier No. 2004663S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Phospho Enol Pyruvate Carboxylase antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Phospho-enol-Pyruvate-Carboxylase [Maize]. Cross reactivity against Phospho-enol-Pyruvate-Carboxylase from other sources may occur but have not been specifically determined.

Phosphoenolpyruvate Carboxylase, through the carboxylation of phosphoenolpyruvate (PEP), forms an oxaloacetate, a four-carbon dicarboxylic acid source for the tricarboxylic acid cycle. Phospho-enol-pyruvate Carboxylase enzyme is regulated by light-reversible phosphorylation. This protein is involved in the pathway C3 acid pathway, which is part of Photosynthesis.

Specifications

Antigen Phospho Enol Pyruvate Carboxylase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Phospho-enol-pyruvate Carboxylase [Maize Leaves]
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.