Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphA2/EphA3/EphA4 (Tyr588, Tyr596) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537811 Supplier Invitrogen™ Supplier No. PA537811
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is HepG2 cells; suggested positive control for ICC/IF is Hela cells.

The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA2/EphA3/EphA4 (Tyr588, Tyr596)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHA3
Gene Accession No. P29317, P29319, P29320, P54764, Q03137, Q03145
Gene Alias 2900005C20Rik; AI385584; ARCC2; AW492086; AW545284; Cek4; Cek8; CTPA; CTPP1; CTRCT6; ECK; EK4; EK8; End3; EPH receptor A2; Eph receptor A3; EPH receptor A4; EPHA2; EPHA3; EPHA4; EPH-like kinase 4; EPH-like kinase 8; eph-like tyrosine kinase 1; Ephrin A2; Ephrin A3; Ephrin A4; ephrin receptor EphA2; ephrin type-A receptor 2; Ephrin type-A receptor 3; ephrin type-A receptor 4; EphrinA2; EphrinA3; EphrinA4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; ETK; ETK1; HEK; HEK4; Hek8; Human embryo kinase; human embryo kinase 1; Mek4; Myk2; rb; receptor protein-tyrosine kinase HEK8; rEK4; RGD1560587; SEK; Sek1; Sek2; Sek-2; soluble EPHA2 variant 1; testicular tissue protein Li 64; Tyro1; TYRO1 protein tyrosine kinase; Tyro4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO1; tyrosine-protein kinase TYRO4
Gene Symbols EPHA2, EPHA3, EPHA4
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 588/596 (K-T-Y(p)-V-R)/(R-T-Y(p)-V-D) derived from Human EPHA2/3/4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13836, 13837, 13838, 1969, 2042, 2043
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.