Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FAK (Tyr407) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 44650G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-650-G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-650-G Supplier Invitrogen™ Supplier No. 44650G
  • $500.00
    $375.65 / Each of 1

    Save $124.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Phospho-FAK (Tyr407) Polyclonal Antibody for Western Blot, IHC

Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of integrin signaling. FAK plays an important role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase transition of the cell cycle. FAK has a central catalytic domain and a C-terminal tail that localizes it to focal adhesions, which are sites where cells attach to the extracellular matrix via surface integrin receptors. Increased FAK tyrosine phosphorylation occurs upon integrin engagement with fibronectin. Adhesion of murine NIH3T3 fibroblasts to fibronectin promotes association of the Grb2 adapter protein and c-Src PTK with FAK in vivo, and also results in activation of the ERK2 MAP kinase. In v-Src-transformed NIH3T3, the association of v-Src, Grb2, and Sos with FAK is independent of cell adhesion to fibronectin. In vitro the Grb2 SH2 domain binds directly to tyrosine-phosphorylated FAK, and the binding site has been identified as Tyr925 by site directed mutagenesis. Several transcript variants encoding different isoforms have been found for the FAK gene, but the full-length natures of only three of them have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FAK (Tyr407)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene PTK2
Gene Accession No. O35346, P34152, Q00944, Q05397
Gene Alias CG10023; CG10023-PA; CG10023-PB; CG10023-PC; CG10023-PD; CG10023-PF; CG10023-PG; CT28129; DFAK; DFak56; Dmel\CG10023; Dmel_CG10023; DmFAK; EC 2.7.10.2; Fadk; FADK 1; Fak; Fak1; Fak56; fak56D; FAK65D; Fak-PA; Fak-PB; Fak-PC; Fak-PD; Fak-PF; Fak-PG; FAK-related non-kinase polypeptide; focal adhesion kinase; Focal adhesion kinase 1; focal adhesion kinase homolog; focal adhesion kinase isoform FAK-Del33; focal adhesion kinase pp125FAK; focal adhesion kinase-related nonkinase; focal ashension kinase 1; FRNK; I79_019430; Kiaa4203; mKIAA4203; p125FAK; p41/p43FRNK; pFAK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein phosphatase 1, regulatory subunit 71; protein tyrosine kinase 2; protein tyrosine kinase 2 L homeolog; protein-tyrosine kinase; protein-tyrosine kinase 2; PTK2; PTK2 protein tyrosine kinase 2; ptk2.L; tyrosine kinase; XELAEV_18032408mg; XPFAK
Gene Symbols PTK2
Host Species Rabbit
Immunogen Synthetic phosphopeptide from human FAK containing tyrosine 407. The sequence is conserved in mouse, rat and chicken.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14083, 25614, 396416, 5747
Target Species Human, Mouse, Rat, Chicken
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
How does the phosphorylation of FAK affect its function?

Focal adhesion kinase (FAK) can be phosphorylated at different sites, this affects its functions as follows:
FAK [pSer722]: Activated in response to cell adhesion; thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pSer732]: This is the site phosphorylated by CDK5. The functional importance is unknown but may play a role in cell cycle regulation in the brain, in particular.
FAK [pSer843]: Activated via cell adhesion, and during mitosis it is thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pSer910]: Activated via cell adhesion, and during mitosis it is thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pTyr397]: Autophosphorylation site (initial activation step) allows Src & p85 subunit of PI3-kinase to bind, and activates cell migration and invasion.
FAK [pTyr407]: Site is activated by TGFbeta and the Epithelial-Mesenchyme Transition (EMT); unlike other sites on FAK, phosphorylation of Tyr407 is EGFR independent.
FAK [pTyr576]: This site is phosphorylated by Src, and is located in the kinase activation loop (YY 576/577 motif); these sites appear to be differentially regulated by stimuli.
FAK [pTyr577]: This site is phosphorylated by Src, and is located in kinase activation loop (YY 576/577 motif), these sites appear to be differentially regulated by stimuli.
FAK [pY861]: This is also a Src site (major), it promotes binding to the cytoplasmic tail of integrins.
FAK [pY925]: This is a Src site (weak), it is GRB2 binding site allowing activation of the Ras/Raf/MEK signaling pathway.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.