Learn More
Invitrogen™ Phospho-FAK (Tyr576, Tyr577) Polyclonal Antibody

Description
A suggested positive control for Western blot is HT29 cells; suggested positive control for ICC/IF is Hela cells.
Specifications
Specifications
| Antigen | Phospho-FAK (Tyr576, Tyr577) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PTK2 |
| Gene Accession No. | O35346, P34152, Q05397 |
| Gene Alias | CG10023; CG10023-PA; CG10023-PB; CG10023-PC; CG10023-PD; CG10023-PF; CG10023-PG; CT28129; DFAK; DFak56; Dmel\CG10023; Dmel_CG10023; DmFAK; EC 2.7.10.2; Fadk; FADK 1; Fak; Fak1; Fak56; fak56D; FAK65D; Fak-PA; Fak-PB; Fak-PC; Fak-PD; Fak-PF; Fak-PG; FAK-related non-kinase polypeptide; focal adhesion kinase; Focal adhesion kinase 1; focal adhesion kinase homolog; focal adhesion kinase isoform FAK-Del33; focal adhesion kinase pp125FAK; focal adhesion kinase-related nonkinase; focal ashension kinase 1; FRNK; I79_019430; Kiaa4203; mKIAA4203; p125FAK; p41/p43FRNK; pFAK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein phosphatase 1, regulatory subunit 71; protein tyrosine kinase 2; protein tyrosine kinase 2 L homeolog; protein-tyrosine kinase; protein-tyrosine kinase 2; PTK2; PTK2 protein tyrosine kinase 2; ptk2.L; tyrosine kinase; XELAEV_18032408mg; XPFAK |
| Gene Symbols | PTK2 |
| Show More |
Frequently Asked Questions (FAQs)
Focal adhesion kinase (FAK) can be phosphorylated at different sites, this affects its functions as follows:
FAK [pSer722]: Activated in response to cell adhesion; thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pSer732]: This is the site phosphorylated by CDK5. The functional importance is unknown but may play a role in cell cycle regulation in the brain, in particular.
FAK [pSer843]: Activated via cell adhesion, and during mitosis it is thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pSer910]: Activated via cell adhesion, and during mitosis it is thought to function in opposition to multiple tyrosine phosphorylation events on FAK.
FAK [pTyr397]: Autophosphorylation site (initial activation step) allows Src & p85 subunit of PI3-kinase to bind, and activates cell migration and invasion.
FAK [pTyr407]: Site is activated by TGFbeta and the Epithelial-Mesenchyme Transition (EMT); unlike other sites on FAK, phosphorylation of Tyr407 is EGFR independent.
FAK [pTyr576]: This site is phosphorylated by Src, and is located in the kinase activation loop (YY 576/577 motif); these sites appear to be differentially regulated by stimuli.
FAK [pTyr577]: This site is phosphorylated by Src, and is located in kinase activation loop (YY 576/577 motif), these sites appear to be differentially regulated by stimuli.
FAK [pY861]: This is also a Src site (major), it promotes binding to the cytoplasmic tail of integrins.
FAK [pY925]: This is a Src site (weak), it is GRB2 binding site allowing activation of the Ras/Raf/MEK signaling pathway.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.