Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FUNDC1 (Ser17) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114576 Supplier Invitrogen™ Supplier No. PA5114576
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of FUNDC1 only when phosphorylated at Serine 17.

Acts as an activator of hypoxia-induced mitophagy, an important mechanism for mitochondrial quality control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FUNDC1 (Ser17)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FUNDC1
Gene Accession No. Q8IVP5
Gene Alias 1500005J14Rik; 1810033P05Rik; FUN14 domain containing 1; FUN14 domain-containing protein 1; fundc1; hypothetical protein LOC436984; si:ch211-222m21.2; zgc:92600
Gene Symbols FUNDC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FUNDC1(Accession Q8IVP5), corresponding to amino acid residues around phosphorylated Ser17.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139341
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.