Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GAB1 (Tyr659) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536908 Supplier Invitrogen™ Supplier No. PA536908
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 76 and 110 kDa. Purity is >95% by SDS-PAGE.

Gab1 is a 115 kDa multiple docking protein that plays an essential role in cellular growth, transformation and apoptosis. Gab1 can be phosphorylated by multiple receptor tyrosine kinase (RTKs), including: insulin receptor (IR), platelet-derived growth factor receptor beta] (PDGFRbeta]), hepatocyte growth factor/scatter factor receptor (HGFR/SFR or c-Met), and epidermal growth factor receptor (EGF), as well as in response to cell-cell adhesion. Gab1 is tyrosine phosphorylated on at least 16 sites, some of which serve as binding sites for phosphoatidylinositol 3-kinase (PI3K), Grb2, PLC gamma 1, Nck, and SHP2. Phosphorylation of Gab1 on tyrosines 627 and 659 is critical for its binding to SHP2, and for activation of the ERK/MAPK pathway in response to EGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GAB1 (Tyr659)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GAB1
Gene Accession No. Q13480, Q9QYY0
Gene Alias AA408973; AW107238; EGK_16125; GAB1; GRB2 associated binding protein 1; GRB2-associated binder 1; GRB2-associated binding protein 1; GRB2-associated binding protein 1 long isoform; GRB2-associated-binding protein 1; Growth factor receptor bound protein 2-associated protein 1; OTTHUMP00000220263
Gene Symbols GAB1
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human Gab 1 around the phosphorylation site of Tyrosine 659.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14388, 2549, 361388
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.