Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GluR2 (Ser880) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104897 Supplier Invitrogen™ Supplier No. PA5104897
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GluR2 only when phosphorylated at Serine 880.

The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GluR2 (Ser880)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gria2
Gene Accession No. P19491, P23819, P42262
Gene Alias AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; Glur2; GluR-2; GLURB; GluR-B; GluR-K2; glutamate ionotropic receptor AMPA type subunit 2; glutamate receptor; glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA2 (alpha 2); GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943
Gene Symbols Gria2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRIA2(Accession P42262), corresponding to amino acid residues around phosphorylated Ser880.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14800, 2891, 29627
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.