Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118740 Supplier Invitrogen™ Supplier No. PA5118740
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GRK1/2 only when phosphorylated at Tyr13.

The sensation of sight is the result of a cascade of events starting with the interaction of photoactivated rhodopsin with a protein called transducin. Rhodopsin kinase is a G-protein-coupled Ser/Thr kinase, also known as GRK1, which is a key element in the regulation of this cascade. Following phosphorylation by GRK1, arrestin is recruited to phospho-rhodopsin quenching its phototransductive activity by preventing further interaction with transducin. By breaking the cycle of phototransduction, GRK1 plays an important role in the restoration of the system for subsequent visual events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GRK1/GRK2 (Tyr13)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRK2
Gene Accession No. P25098, P26817, Q15835, Q63651, Q99MK8, Q9WVL4
Gene Alias ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; G protein-coupled receptor kinase 1; G protein-coupled receptor kinase 2; G protein-coupled receptpr kinase 1; GPRK1; G-protein coupled receptor kinase 2; G-protein receptor kinase 1; G-protein-coupled receptor kinase 2; grk 2; GRK1; Grk2; GRK-2; rhodopsin kinase; Rhodopsin kinase GRK1; Rhok; Rk
Gene Symbols GRK1, GRK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRK2(Accession P25098), corresponding to amino acid residues around phosphorylated Tyr13.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110355, 156, 24013, 25238, 6011, 81760
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.