Learn More
Invitrogen™ Phospho-GRK2 (Ser670) Polyclonal Antibody

Description
This antibody is predicted to react with mouse, rat, hamster and cow.
Specifications
Specifications
| Antigen | Phospho-GRK2 (Ser670) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 |
| Gene | GRK2 |
| Gene Accession No. | P25098 |
| Gene Alias | ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; EC 2.7.1.126; G protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2; Grk2; GRK-2 |
| Gene Symbols | GRK2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.