Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GSK3 alpha/beta (Tyr279, Tyr216) Monoclonal Antibody (M132)
GREENER_CHOICE

Catalog No. PIMA547628
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547628 Supplier Invitrogen™ Supplier No. MA547628
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects 46/50 kDa* proteins corresponding to the apparent molecular mass of GSK-3beta and GSK-3alpha on SDS-PAGE immunoblots of pervanadate treated rabbit fibroblasts, as well as treated human SKN-SH and A431 cells.

Glycogen synthase kinase-3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. Since then, it has been shown to be involved in the regulation of a diverse array of cellular functions, including protein synthesis, cell proliferation, cell differentiation, microtubule assembly/disassembly, and apoptosis. GSK3's substrate specificity is unique in that phosphorylation of the substrate only occurs if a phosphoserine or phosphotyrosine is present - four residues C-terminal to the site of GSK phosphorylation. There exists two isoforms of GSK3, GSK3 alpha and GSK3 beta, and they are strictly regulated via phosphorylation. Phosphorylation of GSK3 beta on Ser9 (Ser21 in GSK3alpha) by protein kinase B (PKB) causes its inactivation and is the primary mechanism responsible for growth factor inhibition of this kinase. Activation of GSK3 beta is dependent upon the phosphorylation of Tyr216 (Tyr279 in GSK3 alpha). Upon activation, it has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3 beta also has been shown to phosphorylate aberrant sites on the microtubule associated protein tau, which is critical for the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GSK3 alpha/beta (Tyr279, Tyr216)
Applications Western Blot
Classification Monoclonal
Clone M132
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene GSK3A
Gene Accession No. P18265, P18266, P49840, P49841, Q2NL51, Q9WV60
Gene Alias 2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B
Gene Symbols GSK3A, GSK3B
Host Species Mouse
Immunogen Clone M132 was generated from a phospho-GSK-3beta (Tyr-216) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 216 of human GSK-3beta. This peptide sequence is also found in GSK-3alpha (Tyr-279) and is highly conserved in GSK-3 genes in rat and mouse.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2931, 2932, 50686, 56637, 606496, 84027
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.