Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HCLS1 (Tyr397) Recombinant Rabbit Monoclonal Antibody (HS1Y397-F12), FITC, Invitrogen™

Catalog No. p-7215079
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537408 10 Tests
PIMA537161 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537408 Supplier Invitrogen™ Supplier No. MA537408
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HCLS1 (Tyr397)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone HS1Y397-F12
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene HCLS1
Gene Accession No. P14317, P49710
Gene Alias AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75
Gene Symbols HCLS1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr397 of human phospho HS1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15163, 3059
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.