Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Histone H1 (Ser1) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540150 Supplier Invitrogen™ Supplier No. PA540150
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Histone H1 (Ser1)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Whole serum with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H1-5
Gene Accession No. D3ZBN0, D4A3K5, P07305, P10922, P15864, P16401, P16403, P43275, P43276, P43278, Q02539
Gene Alias 0610008C09Rik; CTL-1; H1; H1 antibody; H1 histone family member 0; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.0, H1(0), H1-0; H1.1; H1.12 linker histone, cluster member; H1.2; H1.2 linker histone, cluster member; H1.5; H1.9; H10; H1-0; H1-1; H1-12; H1-2; H1A; H1B; H1c; H1F0; H1f1; H1F2; H1F3; H1F5; H1FV; H1s-1; H1s-3; H1var1; H1var3; Hils1; His1a; HIST1; HIST1H1A; HIST1H1B; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1.1; histone H1.2; Histone H1.3; Histone H1.5; histone H1a; Histone H1b; histone H1c; Histone H1d; histone H1-like protein in spermatids 1; histone H1s-1; Histone H1s-2; Histone H1s-3; histone linker H1 domain, spermatid-specific 1 (pseudogene); spermatid-specific linker histone H1-like protein; TISP64
Gene Symbols H1-0, H1-1, H1-2, H1-5
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of serine 1(M-S(p)-E-T) derived from Human Histone H1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116883558, 14958, 24437, 291145, 3005, 3006, 3009, 3024, 50708, 56702, 680522, 80838
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.