Learn More
Invitrogen™ Phospho-Histone H2A.X (Ser139) Monoclonal Antibody (GT2311)
Shop All Thermo Scientific Products

Description
Positive Control: HCT116 (30μM cisplatin treatment for 24hr), NIH-3T3 (30μM cisplatin treatment for 24hr), PC-12 (30uM cisplatin treatment for 24h), U2OS treated UV (50 J/m2) Predicted Reactivity: Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-Histone H2A.X (Ser139) |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT2311 |
| Concentration | 0.92 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | H2AX |
| Gene Accession No. | P16104, P27661 |
| Gene Alias | AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.