Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Phospho-HSP27 (S78/S82) DuoSet IC ELISA

Catalog No. p-4754636
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
15510
24471
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC23142 2 96-well Plates 15510
DYC23145 5 96-well Plates 24471
DYC2314E 15 96-well Plates 15510
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC23142 Supplier R&D Systems™ Supplier No. DYC23142
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Phospho-HSP27 (S78/S82) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Phospho-HSP27 (S78/S82) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat HSP27.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human, Mouse, Rat
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 15510
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.