Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-IGF1R beta (Tyr1135, Tyr1136) Recombinant Rabbit Monoclonal Antibody (15H10L8)

Catalog No. PI701067 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI701067 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI701067 Supplier Invitrogen™ Supplier No. 701067
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The alpha-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IGF1R beta (Tyr1135, Tyr1136)
Applications Western Blot
Classification Recombinant Monoclonal
Clone 15H10L8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene IGF1R
Gene Accession No. P08069
Gene Alias A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acids 1131-1140 of human IGF1R.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.