Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-IkB beta (Thr19, Ser23) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517517 Supplier Invitrogen™ Supplier No. PA517517
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IkB beta (Thr19, Ser23)
Applications Western Blot
Classification Polyclonal
Concentration 8 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene NFKBIB
Gene Accession No. Q15653
Gene Alias I kappa B beta; I(Kappa)B(beta); ikappaBbeta; I-kappa-B-beta; IkB; IKB B; IKB beta; Ikbb; ikB-B; IkBbeta; ikB-beta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; NFKB inhibitor beta; NFKBIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP9; TRIP-9
Gene Symbols NFKBIB
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues of human IkB beta surrounding pThr19/Ser23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4793
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.