Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-JAK1 (Tyr1022/1023), Rabbit, Unlabeled, Polyclonal,

Catalog No. 07849I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07849I 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07849I Supplier MilliporeSigma™ Supplier No. 07849I
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Merlin (UniProt P35240; also known as Moesin-ezrin-radixin-like protein, Neurofibromin-2, Schwannomerlin, Schwannomin) is encoded by the NF2 (also known as SCH) gene (Gene ID 4771) in human. Originally identified as a tumor suppressor gene in the familial Neurofibromatosis type 2 (NF2) cancer patients, NF2 encodes the FERM (Ezrin, Radixin, Moesin) domain-containing merlin that plays a negative regulating role against cell growth and proliferation by functioning as an upstream activator of the Hippo pathway. Merlin directly activates Mst1/2 and recruits Lats1/2 to the plasma membrane for phosphorylation by Mst1/2. The phosphorylated Lats1/2 in turn inhibits YAP/TAZ transcription activity by phosphorylating YAP/TAZ and causes its cytoplasmic sequestration. Merlin also translocates to the nucleus, where it inhibits E3 ubiquitin ligase CRL4(DCAF1), thereby preventing it from ubiquitinating and downregulating Lats1/2. In its inactive state, the C-terminal tail domain (CTD; a.a. 507-595) of merlin is bound intramolecularly to its FERM domain (a.a. 19-313). Angiomotin (AMOT) induces merlin activation by binding to an elongated helical fragment within merlin CTD and subsequently releases the auto-inhibited conformation of Merlin. Merlin CTD can also undergo phosphorylation on Ser518 by p21-activated kinase (PAK) or cyclic AMP-dependent protein kinase (PKA), resulting in a weakened AMOT binding and a stabilization of merlin in its auto-inhibited state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-JAK1 (Tyr1022/1023)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene JAK1, JAK1A, JAK1B
Gene Accession No. P23458
Host Species Rabbit
Immunogen KLH-conjugated phosphopeptide corresponding to the epitope reqeion sequence containing the two phosphorylated target tyrosine residues.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002218
Test Specificity The Tyr1022/1023 designation is according to the JAK1 sequence originally published by Wilks, A.F., et al. (1991). Mol. Cell. Biol. 11(4):2057-2065 (PMID 1848670; NCBI accession: AAA36527). Numbering of the target pTyr residues varies depending on the species and source of the sequence. For example, target pTyr residues this rabbit polyclonal detects correspond to pTyr1034/pTyr1035 and pTyr1033/pTyr1034, respectively, of the human and mouse JAK1 sequence reported by UniProt (P23458 and P52332, respectively), or pTyr1033/pTyr1034 of bovine, canine, and rat JAK1 (NP_001193463 and NP_445918.1), pTyr1021/pTyr1022 of porcine JAK1 (NP_999279).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.