Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Jak1 (Tyr1034, Tyr1035) Recombinant Rabbit Monoclonal Antibody (Jak1Y10341035-F11), PE
GREENER_CHOICE

Catalog No. PIMA536891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536891 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536891 Supplier Invitrogen™ Supplier No. MA536891
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-Jak1 (Tyr1034, Tyr1035) Recombinant Monoclonal Antibody for Flow

Janus kinase 1 (JAK1), is a protein tyrosine kinase that is involved in the interferon-alpha/beta and -gamma signal transduction pathways. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK1. JAK1 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to cis elements upstream of target gene promoters to regulate transcription. Upon ligand binding, JAK1 undergoes tyrosine phosphorylation and catalytic activation in an interdependent manner. Phosphorylation of tyrosine residues at positions 1022 and 1023 is believed to function in the activation of catalytic events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Jak1 (Tyr1034, Tyr1035)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone Jak1Y10341035-F11
Conjugate PE
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene JAK1
Gene Accession No. P23458, P52332
Gene Alias AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1
Gene Symbols JAK1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr1034/1035 of human phospho-Jak1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16451, 3716
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.