Learn More
Invitrogen™ Phospho-JAK1 (Tyr1034, Tyr1035) Recombinant Rabbit Monoclonal Antibody (59H4L5)
Description
This antibody is predicted to react with mouse, rat, chimpanzee, Rhesus monkey, bovine, canine, porcine, equine, Xenopus and zebrafish based on sequence homology.Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain.Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.This product was formerly called Phospho-JAK1 (Tyr1022, Tyr1023) because Human Jak1 residues Tyr1034 and Tyr1035 have been historically referenced as Tyr1022 and Tyr1023.
Specifications
Specifications
| Antigen | Phospho-JAK1 (Tyr1034, Tyr1035) |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 59H4L5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | JAK1 |
| Gene Accession No. | P23458, P52332 |
| Gene Alias | AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.