Learn More
Invitrogen™ Phospho-JNK1/JNK2 (Thr183, Tyr185) Monoclonal Antibody (E.665.10)
Shop All Thermo Scientific Products

Description
Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay. It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | Phospho-JNK1/JNK2 (Thr183, Tyr185) |
| Applications | ChIP Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | E.665.10 |
| Concentration | 591 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100μg/mL BSA and <0.02% sodium azide; pH 7.5 |
| Gene | MAPK9 |
| Gene Accession No. | P45983, P45984, P49185, P49186, Q91Y86, Q9WTU6 |
| Gene Alias | AI849689; AI851083; C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; Mapk8; Mapk9; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; OTTHUMP00000019552; p54 gamma; p54a; p54-alpha; p54aSAPK; Phospho-JNK; PRKM; PRKM8; Prkm9; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK; SAPK 1; SAPK gamma; SAPK1; SAPK1A; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1c; stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.