Learn More
Invitrogen™ Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody

Description
PA1-9593 detects Phospho-JNK/SAPK in human, mouse, and rat samples. PA1-9593 has been successfully used in Western blot, immunohistochemistry, and immunocytochemistry procedures. By Western blot, PA1-9593 detects a 46 kDa and 54 kDa protein representing Phospho-JNK/SAPK. PA1-9593 immunogen is a phosphorylated peptide (pT183-P-pY185) based on human JNK.
Specifications
Specifications
| Antigen | Phospho-JNK1/JNK2 (Thr183, Tyr185) |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | MAPK9 |
| Gene Accession No. | P45983, P45984, P49185, P49186, Q91Y86, Q9WTU6 |
| Gene Alias | AI849689; AI851083; C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; Mapk8; Mapk9; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; OTTHUMP00000019552; p54 gamma; p54a; p54-alpha; p54aSAPK; Phospho-JNK; PRKM; PRKM8; Prkm9; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK; SAPK 1; SAPK gamma; SAPK1; SAPK1A; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1c; stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2 |
| Gene Symbols | MAPK8, MAPK9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.