Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-KCC2 (Thr1007) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595677 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595677 Supplier Invitrogen™ Supplier No. PA595677
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antigen Affinity Purified from Pooled Serum.

Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-KCC2 (Thr1007)
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene SLC12A5
Gene Accession No. Q63633, Q91V14
Gene Alias EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5
Gene Symbols SLC12A5
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr1007 of rat KCC2 conjugated to KLH.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171373, 57138
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.