Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-LRP6 (Thr1479) Polyclonal Antibody, Abbomax

Catalog No. 502203954
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3954 Supplier Abbomax Inc Supplier No. 601060
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human breast cancer Cellular location: Cytoplasm, Nucleus.

Phospho-LRP6 (Thr1479) Polyclonal antibody specifically detects Phospho-LRP6 (Thr1479) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Phospho-LRP6 (Thr1479)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene LRP6
Gene Accession No. O75581, O88572
Gene Alias ADCAD2; C030016K15Rik; Cd; crooked; FLJ90062; FLJ90421; Gw; LDL receptor related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; low-density lipoprotein receptor-related protein 6; lpr6; LRP6; LRP-6; ska<m26Jus>; ska26; skax26; STHAG7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the epitope KGTYF at a phosphorylation threonine1479 in LRP6 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16974, 312781, 4040
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.