Learn More
Cell Signaling Technology Phospho-LYN (Tyr397)/LCK (Tyr394)/HCK (Tyr411)/BLK (Tyr389) (E5L3D) Rabbit mAb (Alexa Fluor(R) 647 Conjugate) 100 ul
SDP
Supplier:  Cell Signaling Technology 78740S
Encompass
Phospho-LYN (Tyr397)/LCK (Tyr394)/HCK (Tyr411)/BLK (Tyr389) (E5L3D) Rabbit mAb (Alexa Fluor(R) 647 Conjugate) 100 ul
Catalog No. 50-273-1260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.