Learn More
Invitrogen™ Phospho-MAPKAPK2 (Thr334) Recombinant Rabbit Monoclonal Antibody (MAPKAPK2T334-H2), PE, Invitrogen™
Description
Specifications
Specifications
| Antigen | Phospho-MAPKAPK2 (Thr334) |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | MAPKAPK2T334-H2 |
| Conjugate | PE |
| Formulation | PBS with 0.20% BSA and 0.09% sodium azide |
| Gene | MAPKAPK2 |
| Gene Accession No. | P49137, P49138 |
| Gene Alias | AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1 |
| Gene Symbols | MAPKAPK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.