Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517495 Supplier Invitrogen™ Supplier No. PA517495
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

This antibody does not react with MARK4. It is not recommended to aliquot this antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234)
Applications Western Blot
Classification Polyclonal
Concentration 59 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 0.15M NaCl, 50% glycerol and no preservative; pH 7.5
Gene MARK3
Gene Accession No. O08678, O08679, P27448, Q03141, Q05512, Q7KZI7, Q8VHF0, Q8VHJ5, Q9P0L2
Gene Alias 1600015G02Rik; A430080F22Rik; AW491150; B930025N23Rik; cdc25C-associated protein kinase 1; CTAK1; C-TAK1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk; EMK1; EMK-1; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; Emk3; ETK1; ETK-1; Kiaa1477; KP78; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity regulating kinase 2; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 3; MARK; MARK1; mark1 {ECO:0000312; MARK2; mark2 {ECO:0000312; MARK3; microtubule affinity regulating kinase 1; microtubule affinity regulating kinase 2; microtubule affinity regulating kinase 3; mKIAA1477; mPar-1c; MPK10; MPK-10; Par-1; PAR1 homolog; PAR1 homolog b; PAR1 homolog c; PAR1A; Par-1a; Par1b; Par-1b; Par1c; Par-1c; Protein kinase STK10; RGD:619882}; RGD:708483}; ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase MARK1; Serine/threonine-protein kinase MARK2; serine/threonine-protein kinase p78; testicular tissue protein Li 117
Gene Symbols MARK1, MARK2, MARK3
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding threonine 215 of human MARK1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117016, 13728, 170577, 17169, 2011, 226778, 4139, 4140, 60328
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.