Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MEF2A (Thr319) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537643 Supplier Invitrogen™ Supplier No. PA537643
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for ICC/IF is Hela cells.

The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MEF2A (Thr319)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MEF2A
Gene Accession No. Q02078, Q2MJT0, Q60929
Gene Alias A430079H05Rik; ADCAD1; C5DELq14.3; DEL5q14.3; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); MADS box transcription enhancer factor 2, polypeptide C; mef2; Mef2a; MEF2C; myocyte enhancer factor 2a; myocyte enhancer factor 2C; Myocyte-specific enhancer factor 2A; myocyte-specific enhancer factor 2C; RSRFC4; RSRFC9; Serum response factor-like protein 1
Gene Symbols MEF2A
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Thr319 (V-T-T(p)-P-S) derived from Human MEF2A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17258, 309957, 4205
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.