Learn More
Invitrogen™ Phospho-MEK3/MEK6 (Ser189, Ser207) Recombinant Rabbit Monoclonal Antibody (MKK3S189MKK6S207-D3), FITC, Invitrogen™
Description
Specifications
Specifications
| Antigen | Phospho-MEK3/MEK6 (Ser189, Ser207) |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | MKK3S189MKK6S207-D3 |
| Conjugate | FITC |
| Formulation | PBS with 0.20% BSA and 0.09% sodium azide |
| Gene | MAP2K3 |
| Gene Accession No. | O09110, P46734, P52564, P70236 |
| Gene Alias | AW212142; dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; Map2k3; Map2k6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 3; mitogen-activated protein kinase kinase 6; MKK3; MKK6; mMKK3b; OTTHUMP00000166044; Prkmk3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK2; SKK3; Stress-activated protein kinase kinase 2; stress-activated protein kinase kinase 3 |
| Gene Symbols | MAP2K3, MAP2K6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.