Learn More
Invitrogen™ Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-MER/SKY (Tyr749, Tyr681) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | MERTK |
| Gene Accession No. | P55144, P55146, P57097, Q06418, Q12866, Q60805 |
| Gene Alias | AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Dtk; EC 2.7.10.2; Etk-2; etk2/tyro3; Eyk; kinase Syk; KSYK; Mer; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; Receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; Rek; retinal dystrophy; RP38; RSE; Sky; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; SYK; Tif; TK19-2; Tyro 12; Tyro12; Tyro3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase Mer; Tyrosine-protein kinase receptor TYRO3; tyrosine-protein kinase RSE; Tyrosine-protein kinase SKY; tyrosine-protein kinase TIF |
| Gene Symbols | MERTK, TYRO3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.