Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MSK1 (Thr581) Recombinant Rabbit Monoclonal Antibody (MSK1T581-A5), FITC
GREENER_CHOICE

Catalog No. PIMA536890
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536890 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536890 Supplier Invitrogen™ Supplier No. MA536890
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-MSK1 (Thr581) Recombinant Monoclonal Antibody for Flow

RPS6KA5 (MSK1) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes. RPS6KA5 is required for the mitogen or stress-induced phosphorylation of the transcription factors CREB1 and ATF1 and for the regulation of the transcription factors RELA, STAT3 and ETV1/ER81, and that contributes to gene activation by histone phosphorylation and functions in the regulation of inflammatory genes. Phosphorylates CREB1 and ATF1 in response to mitogenic or stress stimuli such as UV-C irradiation, epidermal growth factor (EGF) and anisomycin. Plays an essential role in the control of RELA transcriptional activity in response to TNF and upon glucocorticoid, associates in the cytoplasm with the glucocorticoid receptor NR3C1 and contributes to RELA inhibition and repression of inflammatory gene expression. In skeletal myoblasts is required for phosphorylation of RELA at 'Ser-276' during oxidative stress. In erythropoietin-stimulated cells, is necessary for the 'Ser-727' phosphorylation of STAT3 and regulation of its transcriptional potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MSK1 (Thr581)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone MSK1T581-A5
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene Rps6ka5
Gene Accession No. O75582, Q8C050
Gene Alias 3110005L17Rik; 6330404E13Rik; 90 kDa ribosomal protein S6 kinase 5; AI854034; kinase MSK1; KS6A5; mitogen and stress-activated protein kinase 1; Msk1; MSPK1; Nuclear mitogen- and stress-activated protein kinase 1; ribosomal protein S6 kinase A5; ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; ribosomal protein S6 kinase, polypeptide 5; RLPK; RLSK; Rps6ka5; RSKL; RSK-like protein kinase; S6K-alpha-5
Gene Symbols Rps6ka5
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Thr581 of human phospho MSK1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73086, 9252
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.