Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Myb (Ser11) Recombinant Rabbit Monoclonal Antibody (K01_2K06)
GREENER_CHOICE

Catalog No. MA563922
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563922 100 μL
MA565381 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563922 Supplier Invitrogen™ Supplier No. MA563922
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Phospho-Myb (Ser11) Recombinant Monoclonal Antibody for IHC (P), Western Blot

Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Myb (Ser11)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2K06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene MYB
Gene Accession No. P10242
Gene Alias AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Symbols MYB
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser11 of human v-Myb/c-Myb.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4602
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.