Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MYF5 (Ser49) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988361
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8361 Supplier Bioss Supplier No. BS8200R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYF5 is a member of the myogenic basic helix-loop-helix family of transcription factors, which can activate the muscle differentiation program.

Specifications

Antigen Phospho-MYF5 (Ser49)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYF5
Gene Accession No. P13349
Gene Alias B130010J22Rik; basic helix-loop-helix nuclear protein; BHLHC2; BOS_4874; cb641; Class C basic helix-loop-helix protein 2; LOC100351906; muscle regulatory protein Myf5; Myf5; Myf-5; myf5 protein; MyoD family; Myogenic factor 5; myogenic factor 5-like; myogenic factor-5; myogenic regulatory factor 5; myogenic regulatory factor MYF-5; zgc:110509
Gene Symbols MYF5
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MYF5 around the phosphorylation site of Ser49.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4617
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.